siRNA Details
Gene Symbol: Appl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008771023.1 | XP_008769245.1 | 1 | 180 | ||
GenBank mRNA | FQ226179.1 | - | 1 | 180 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Appl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008771023.1 | XP_008769245.1 | 1 | 180 | ||
GenBank mRNA | FQ226179.1 | - | 1 | 180 |