siRNA Details
Gene Symbol: Arhgef3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106061.1 | NP_001099531.1 | 7, 8 | 977 | ||
XM_006252574.2 | XP_006252636.1 | 10, 11 | 1109 | |||
XM_006252575.2 | XP_006252637.1 | 10, 11 | 1307 | |||
XM_006252576.2 | XP_006252638.1 | 8, 9 | 1473 | |||
XM_008770982.1 | XP_008769204.1 | 10, 11 | 1174 | |||
XM_008770983.1 | XP_008769205.1 | 10, 11 | 1016 |
