siRNA Details
Gene Symbol: LOC290595 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106063.2 | NP_001099533.2 | 3 | 381 | ||
GenBank mRNA | AF152002.1 | - | 3 | 384 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC290595 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106063.2 | NP_001099533.2 | 3 | 381 | ||
GenBank mRNA | AF152002.1 | - | 3 | 384 |