siRNA Details
Gene Symbol: Ghitm | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005908.1 | NP_001005908.1 | 9 | 1103 | ||
XM_006252786.2 | XP_006252848.1 | 9 | 1357 | |||
GenBank mRNA | BC083778.1 | - | 9 | 1103 | 7193683 | |
FQ212260.1 | - | 9 | 1303 | |||
FQ217853.1 | - | 9 | 1121 |
