siRNA Details
Gene Symbol: Gatad2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013881.2 | NP_001013903.2 | 8 | 1431 | ||
XM_006252879.2 | XP_006252941.1 | 7 | 1111 | |||
XM_006252880.2 | XP_006252942.1 | 7 | 1108 | |||
XM_006252881.2 | XP_006252943.1 | 7 | 1102 | |||
XM_006252882.2 | XP_006252944.1 | 7 | 1099 | |||
XM_006252883.2 | XP_006252945.1 | 7 | 1111 | |||
XM_006252884.2 | XP_006252946.1 | 8 | 1390 | |||
XM_006252887.2 | XP_006252949.1 | 7 | 923 | |||
XM_006252889.2 | XP_006252951.1 | 7 | 1109 | |||
XM_008771089.1 | XP_008769311.1 | 9 | 1480 | |||
XM_008771090.1 | XP_008769312.1 | 8 | 1110 | |||
GenBank mRNA | BC089902.1 | - | 5 | 521 | 7383529 |
