siRNA Details
Gene Symbol: Trappc11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001169115.1 | NP_001162586.1 | 21 | 2568 | ||
XM_006253134.2 | XP_006253196.1 | 21 | 2489 | |||
XM_006253135.2 | XP_006253197.1 | 21 | 2424 |
