siRNA Details
Gene Symbol: F11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001047848.1 | NP_001041313.1 | 7, 8 | 834 | ||
XM_006253144.2 | XP_006253206.1 | 8, 9 | 1136 | |||
XM_006253145.1 | XP_006253207.1 | 6, 7 | 745 | |||
GenBank mRNA | AY387072.1 | - | 7, 8 | 834 |
