siRNA Details
Gene Symbol: Tusc3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004212.1 | NP_001004212.1 | 1 | 249 | ||
XM_008771294.1 | XP_008769516.1 | 1 | 267 | |||
GenBank mRNA | BC079193.1 | - | 1 | 249 | 7131345 | |
FQ213307.1 | - | 1 | 267 |
