siRNA Details
Gene Symbol: Tex15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106087.1 | NP_001099557.1 | 1 | 6427 | ||
XM_006253218.2 | XP_006253280.1 | 8 | 7471 | |||
XM_006253221.2 | XP_006253283.1 | 5 | 6974 | |||
XM_008771296.1 | XP_008769518.1 | 8 | 7443 | |||
XM_008771297.1 | XP_008769519.1 | 8 | 7487 | |||
XM_008771298.1 | XP_008769520.1 | 7 | 7311 | |||
XM_008771299.1 | XP_008769521.1 | 4 | 6900 |
