siRNA Details
Gene Symbol: Mrps31 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106091.1 | NP_001099561.1 | 4 | 715 | ||
XM_006253349.2 | XP_006253411.1 | 1936 | ||||
GenBank mRNA | BC158691.1 | - | 4 | 701 | 9021908 |
