siRNA Details
Gene Symbol: LOC100361383 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006253741.2 | XP_006253803.1 | 9 | 1732 | ||
XM_006253742.2 | XP_006253804.1 | 8 | 1543 | |||
GenBank mRNA | FQ215770.1 | - | 3 | 321 |
Gene Symbol: LOC100910978 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006253693.2 | XP_006253755.1 | 3096 | |||
XM_006253694.2 | XP_006253756.1 | 2907 | ||||
XM_008771523.1 | XP_008769745.1 | 1601 |
