siRNA Details
Gene Symbol: Mcur1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_214448.8 | XP_214448.5 | 8 | 1546 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Mcur1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_214448.8 | XP_214448.5 | 8 | 1546 |