siRNA Details
Gene Symbol: Eci2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001006966.1 | NP_001006967.1 | 5 | 580 | ||
GenBank mRNA | BC083764.1 | - | 5 | 580 | 7193133 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Eci2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001006966.1 | NP_001006967.1 | 5 | 580 | ||
GenBank mRNA | BC083764.1 | - | 5 | 580 | 7193133 |