siRNA Details
Gene Symbol: Irf4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106108.1 | NP_001099578.1 | 7 | 823 | ||
XM_006253899.2 | XP_006253961.1 | 7 | 895 | |||
XM_006253900.2 | XP_006253962.1 | 6 | 790 |
