siRNA Details
Gene Symbol: Gmnn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106112.1 | NP_001099582.1 | 4 | 601 | ||
XM_006253927.2 | XP_006253989.1 | 4 | 386 | |||
XM_006253928.2 | XP_006253990.1 | 4 | 324 | |||
GenBank mRNA | FQ219913.1 | - | 1 | 89 |
