siRNA Details
Gene Symbol: Actn2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001170325.1 | NP_001163796.1 | 21 | 2768 | ||
GenBank mRNA | CR461659.1 | - | 1 | 173 | ||
FM123616.1 | - | 5 | 482 | |||
FQ215597.1 | - | 8 | 1048 | |||
FQ224888.1 | - | 7 | 907 |
