siRNA Details
Gene Symbol: Zmynd11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_203366.2 | NP_976242.2 | 10 | 1143 | ||
NM_203367.2 | NP_976243.2 | 10 | 1143 | |||
NM_203368.2 | NP_976244.2 | 9 | 981 | |||
NM_203369.2 | NP_976245.2 | 10 | 1062 | |||
XM_006254150.2 | XP_006254212.1 | 12 | 1373 | |||
XM_006254151.2 | XP_006254213.1 | 12 | 1426 | |||
XM_006254152.2 | XP_006254214.1 | 12 | 1285 | |||
XM_006254154.2 | XP_006254216.1 | 11 | 1328 | |||
XM_006254155.2 | XP_006254217.1 | 11 | 1380 | |||
XM_008771704.1 | XP_008769926.1 | 12 | 1423 | |||
XM_008771705.1 | XP_008769927.1 | 12 | 1424 | |||
XM_008771706.1 | XP_008769928.1 | 10 | 1116 | |||
XM_008771707.1 | XP_008769929.1 | 10 | 1218 | |||
XM_008771708.1 | XP_008769930.1 | 11 | 1378 | |||
XM_008771709.1 | XP_008769931.1 | 11 | 1376 | |||
XM_008771710.1 | XP_008769932.1 | 10 | 1216 | |||
XM_008771711.1 | XP_008769933.1 | 10 | 1214 | |||
GenBank mRNA | AB162617.1 | - | 9 | 1004 | ||
AB162618.1 | - | 9 | 1004 | |||
AB162619.1 | - | 8 | 842 | |||
AB162620.1 | - | 8 | 842 | |||
BC065308.1 | - | 10 | 1059 | 5599817 |
