siRNA Details
Gene Symbol: Fbxo18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106119.1 | NP_001099589.1 | 18 | 2826 | ||
XM_006254200.2 | XP_006254262.1 | 18 | 2927 | |||
XM_006254201.2 | XP_006254263.1 | 18 | 2953 | |||
XM_006254202.2 | XP_006254264.1 | 18 | 2856 | |||
XM_006254204.2 | XP_006254266.1 | 18 | 2774 |
