siRNA Details
Gene Symbol: Usp6nl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106120.1 | NP_001099590.1 | 7 | 592 | ||
XM_006254232.2 | XP_006254294.1 | 6 | 836 | |||
XM_006254233.1 | XP_006254295.1 | 6 | 461 | |||
XM_006254236.2 | XP_006254298.1 | 4 | 1942 |
