siRNA Details
Gene Symbol: Sephs1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001104630.1 | NP_001098100.1 | 8 | 1288 | ||
XM_006254266.2 | XP_006254328.1 | 8 | 2915 | |||
GenBank mRNA | AY724491.1 | - | 8 | 1288 |
