siRNA Details
Gene Symbol: Atp1a4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271030.1 | NP_001257959.1 | 19 | 2717 | ||
NM_022848.3 | NP_074039.1 | 19 | 2720 | |||
GenBank mRNA | BC087015.1 | - | 19 | 2709 | 7113151 | |
U15176.1 | - | 19 | 2720 |
