siRNA Details
Gene Symbol: Zfp516 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001191695.1 | NP_001178624.1 | 1 | 1360 | ||
XM_008772098.1 | XP_008770320.1 | 1 | 1376 | |||
XM_008772099.1 | XP_008770321.1 | 1 | 1376 | |||
XM_008772100.1 | XP_008770322.1 | 1 | 1376 |
