siRNA Details
Gene Symbol: Cxxc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001079698.1 | NP_001073166.1 | 7 | 926 | ||
XM_006254917.2 | XP_006254979.1 | 7 | 1215 | |||
GenBank mRNA | BC087117.1 | - | 6 | 1166 | ||
BC128781.1 | - | 7 | 926 | 7370019 |
