siRNA Details
Gene Symbol: Megf10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100657.1 | NP_001094127.1 | 23 | 3058 | ||
XM_006254747.1 | XP_006254809.1 | 24 | 3763 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Megf10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100657.1 | NP_001094127.1 | 23 | 3058 | ||
XM_006254747.1 | XP_006254809.1 | 24 | 3763 |