siRNA Details
Gene Symbol: Megf10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100657.1 | NP_001094127.1 | 5 | 650 | ||
XM_006254747.1 | XP_006254809.1 | 6 | 1355 | |||
GenBank mRNA | L41686.1 | - | 1 | 39 |
