siRNA Details
Gene Symbol: Poli | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106137.1 | NP_001099607.1 | 5 | 646 | ||
XM_006254900.2 | XP_006254962.1 | 5 | 567 | |||
XM_006254901.2 | XP_006254963.1 | 4 | 509 | |||
XM_008772110.1 | XP_008770332.1 | 2 | 501 |
