siRNA Details
Gene Symbol: Poli | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106137.1 | NP_001099607.1 | 8 | 1125 | ||
XM_006254900.2 | XP_006254962.1 | 8 | 1046 | |||
XM_006254901.2 | XP_006254963.1 | 7 | 988 | |||
XM_008772110.1 | XP_008770332.1 | 5 | 980 |
