siRNA Details
Gene Symbol: Ndfip1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013059.1 | NP_001013077.1 | 7 | 783 | ||
XM_006254633.2 | XP_006254695.1 | 7 | 851 | |||
GenBank mRNA | BC085887.1 | - | 7 | 783 | 7192378 |
