siRNA Details
Gene Symbol: Dnajc18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013887.1 | NP_001013909.1 | 8 | 994 | ||
XM_006254546.2 | XP_006254608.1 | 7 | 973 | |||
GenBank mRNA | BC079289.1 | - | 8 | 994 | 7134364 | |
DQ158861.1 | - | 7 | 927 | |||
FQ214241.1 | - | 7 | 847 |
