siRNA Details
Gene Symbol: Dctn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024130.1 | NP_077044.1 | 29 | 3781 | ||
GenBank mRNA | FQ211585.1 | - | 7 | 805 | ||
FQ223809.1 | - | 3486 | ||||
X62160.1 | - | 29 | 3781 |
