siRNA Details
Gene Symbol: LOC103689930 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006254461.2 | XP_006254523.1 | 21 | 3517 | ||
XM_006254462.2 | XP_006254524.1 | 21 | 3514 | |||
XM_008771994.1 | XP_008770216.1 | 21 | 3571 |
Gene Symbol: Trappc8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106160.1 | NP_001099630.1 | 3271 | |||
GenBank mRNA | FQ230517.1 | - | 1128 |
