siRNA Details
Gene Symbol: RGD1562997 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006254439.2 | XP_006254501.1 | 6 | 1272 | ||
XM_006254440.2 | XP_006254502.1 | 5 | 507 | |||
XM_008771989.1 | XP_008770211.1 | 6 | 1272 | |||
XM_008771990.1 | XP_008770212.1 | 6 | 1272 |
