siRNA Details
Gene Symbol: Rbbp8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134417.1 | NP_001127889.1 | 16, 17 | 2613 | ||
XM_006254407.1 | XP_006254469.1 | 16, 17 | 2757 | |||
XM_006254408.2 | XP_006254470.1 | 16, 17 | 2779 | |||
XM_008771938.1 | XP_008770160.1 | 16, 17 | 2836 | |||
XM_008771939.1 | XP_008770161.1 | 16, 17 | 2682 | |||
XM_008771940.1 | XP_008770162.1 | 16, 17 | 2658 | |||
XM_008771941.1 | XP_008770163.1 | 16, 17 | 2734 | |||
XM_008771942.1 | XP_008770164.1 | 16, 17 | 2839 | |||
GenBank mRNA | BC162012.1 | - | 16, 17 | 2613 | 9091712 | |
FQ209715.1 | - | 16, 17 | 2665 |
