siRNA Details
Gene Symbol: Slc38a7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001003705.2 | NP_001003705.1 | 2 | 426 | ||
XM_006255067.2 | XP_006255129.1 | 4 | 573 | |||
GenBank mRNA | AY294328.1 | - | 3 | 521 | ||
BC086369.1 | - | 2 | 426 | 7109456 |
