siRNA Details
Gene Symbol: Prss54 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013888.1 | NP_001013910.1 | 4 | 626 | ||
XM_006255073.2 | XP_006255135.1 | 4 | 642 | |||
XM_008772273.1 | XP_008770495.1 | 4 | 513 | |||
GenBank mRNA | BC079217.1 | - | 4 | 626 | 7131896 |
