siRNA Details
Gene Symbol: Gpr97 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006255126.2 | XP_006255188.1 | 9, 10 | 3185 | ||
XM_006255127.2 | XP_006255189.1 | 8, 9 | 3038 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Gpr97 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006255126.2 | XP_006255188.1 | 9, 10 | 3185 | ||
XM_006255127.2 | XP_006255189.1 | 8, 9 | 3038 |