siRNA Details
Gene Symbol: Kctd19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008772558.1 | XP_008770780.1 | 2 | 224 | ||
XM_226403.8 | XP_226403.6 | 2 | 289 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Kctd19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008772558.1 | XP_008770780.1 | 2 | 224 | ||
XM_226403.8 | XP_226403.6 | 2 | 289 |