siRNA Details
Gene Symbol: Atp6v0d1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001011927.1 | NP_001011927.1 | 7, 8 | 962 | ||
GenBank mRNA | BC088462.1 | - | 7, 8 | 962 | 7127604 | |
FQ213716.1 | - | 7, 8 | 979 | |||
FQ215412.1 | - | 6, 7 | 698 |
