siRNA Details
Gene Symbol: Hfe | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001173434.1 | NP_001166905.1 | 2 | 260 | ||
NM_053301.4 | NP_445753.1 | 3 | 524 | |||
GenBank mRNA | AF176534.1 | - | 2 | 180 | ||
AJ001517.1 | - | 3 | 563 | |||
FQ219597.1 | - | 2 | 274 |
