siRNA Details
Gene Symbol: Dhx38 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106185.1 | NP_001099655.1 | 9 | 1517 | ||
XM_006255577.2 | XP_006255639.1 | 9 | 1563 | |||
XM_008772489.1 | XP_008770711.1 | 9 | 2106 |

More Information
Other Species Targeted

Gene Ontology Categories:
