siRNA Details
Gene Symbol: Aars | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100517.1 | NP_001093987.1 | 2 | 126 | ||
XM_006255580.2 | XP_006255642.1 | 2 | 364 | |||
XM_006255581.2 | XP_006255643.1 | 3 | 221 | |||
XM_006255582.2 | XP_006255644.1 | 2 | 360 | |||
XM_006255583.2 | XP_006255645.1 | 2 | 116 |
