siRNA Details
Gene Symbol: Banp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106191.1 | NP_001099661.1 | 10, 11 | 1406 | ||
XM_006255751.2 | XP_006255813.1 | 10, 11 | 1452 | |||
XM_006255752.2 | XP_006255814.1 | 10, 11 | 1442 | |||
XM_006255753.2 | XP_006255815.1 | 9, 10 | 1334 | |||
XM_006255754.2 | XP_006255816.1 | 9, 10 | 1325 | |||
XM_006255755.2 | XP_006255817.1 | 10, 11 | 1451 | |||
XM_006255756.2 | XP_006255818.1 | 10, 11 | 1442 | |||
XM_006255757.2 | XP_006255819.1 | 9, 10 | 1324 | |||
XM_008772601.1 | XP_008770823.1 | 10, 11 | 1673 | |||
XM_008772602.1 | XP_008770824.1 | 10, 11 | 1452 | |||
GenBank mRNA | BC160844.1 | - | 10, 11 | 1433 | 8361869 |
