siRNA Details
Gene Symbol: Ctu2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037094.1 | NP_001032171.1 | 6 | 416 | ||
GenBank mRNA | BC079393.1 | - | 6 | 466 | ||
BC089862.1 | - | 6 | 414 | |||
BC107464.1 | - | 6 | 416 | 7454431 |
