siRNA Details
Gene Symbol: Ttll2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001169136.1 | NP_001162607.1 | 2 | 894 | ||
XM_006227919.2 | XP_006227981.1 | 3 | 4191 | |||
XM_006227921.2 | XP_006227983.1 | 3 | 4383 | |||
XM_008758751.1 | XP_008756973.1 | 3 | 4194 |
