siRNA Details
Gene Symbol: LOC102554917 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006228360.1 | XP_006228422.1 | 4 | 570 |
Gene Symbol: Obox1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006228359.1 | XP_006228421.1 | 2 | 543 |
