siRNA Details
Gene Symbol: Gpi | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207592.1 | NP_997475.1 | 12, 13 | 1132 | ||
GenBank mRNA | BC062005.1 | - | 12, 13 | 1132 | 5599776 | |
FQ215247.1 | - | 1, 2 | 179 | |||
FQ216077.1 | - | 1, 2 | 180 |
