siRNA Details
Gene Symbol: Cep89 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127599.1 | NP_001121071.1 | 13, 14 | 1432 | ||
XM_006228881.2 | XP_006228943.1 | 13, 14 | 1501 | |||
XM_006228883.2 | XP_006228945.1 | 13, 14 | 1501 | |||
GenBank mRNA | BC161945.1 | - | 13, 14 | 1432 | 9025363 |
