siRNA Details
Gene Symbol: LOC100912350 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003748841.3 | XP_003748889.1 | 1656 | |||
XM_006228898.2 | XP_006228960.1 | 1656 |
Gene Symbol: Zfp507 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106248.1 | NP_001099718.2 | 3 | 1193 | ||
XM_006228885.2 | XP_006228947.1 | 3 | 1154 | |||
XM_006228886.2 | XP_006228948.1 | 3 | 1225 |
