siRNA Details
Gene Symbol: Tjp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106266.1 | NP_001099736.1 | 18 | 2586 | ||
XM_006229330.2 | XP_006229392.1 | 19 | 2729 | |||
XM_006229331.2 | XP_006229393.1 | 19 | 2818 | |||
XM_006229332.2 | XP_006229394.1 | 19 | 3108 | |||
XM_006229333.2 | XP_006229395.1 | 19 | 2628 | |||
XM_008759501.1 | XP_008757723.1 | 19 | 3052 | |||
XM_008759502.1 | XP_008757724.1 | 19 | 3052 | |||
XM_008759503.1 | XP_008757725.1 | 19 | 3052 | |||
XM_008759504.1 | XP_008757726.1 | 19 | 3052 |
