siRNA Details
Gene Symbol: Klhl25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039006.1 | NP_001034095.1 | 2 | 1679 | ||
XM_006229379.2 | XP_006229441.1 | 2 | 1992 | |||
XM_006229380.2 | XP_006229442.1 | 2 | 6030 | |||
XM_006229381.2 | XP_006229443.1 | 2 | 5085 | |||
XM_008759505.1 | XP_008757727.1 | 2 | 1722 | |||
GenBank mRNA | BC099111.1 | - | 2 | 1679 | 7461562 | |
FQ230538.1 | - | 2 | 1687 |
